5-Chloro-L-tryptophan 97%

Supplier: Apollo Scientific

OR452007-1G OR452007-250MG
APOSOR452007-1GEA 441 EUR
APOSOR452007-1G APOSOR452007-250MG
5-Chloro-L-tryptophan 97%
5-Chloro-L-tryptophan
Formula: C₁₁H₁₁ClN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD06797629
CAS Number: 52448-15-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR