Boc-(R)-3-Cyclobutylalanine 97%
Supplier: Apollo Scientific
OR452010-1G
OR452010-250MG
OR452010-500MG
APOSOR452010-1GEA
522
EUR
APOSOR452010-1G
APOSOR452010-250MG
APOSOR452010-500MG
Boc-(R)-3-Cyclobutylalanine 97%
Boc-(R)-3-Cyclobutylalanine
Formula:
C₁₂H₂₁NO₄ MW: 243.299 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03840393 CAS Number: 478183-61-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...