4-(1-Methylethyl)-L-phenylalanine 97%
Supplier: Apollo Scientific
OR452019-1G
OR452019-500MG
APOSOR452019-1GEA
522
EUR
APOSOR452019-1G
APOSOR452019-500MG
4-(1-Methylethyl)-L-phenylalanine 97%
4-(1-Methylethyl)-L-phenylalanine
Formula:
C₁₂H₁₇NO₂ MW: 207,269 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01860653 CAS Number: 216007-00-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...