4-Chloro-DL-homophenylalanine 97%
Supplier: Apollo Scientific
OR452038-1G
OR452038-250MG
OR452038-500MG
APOSOR452038-1GEA
655
EUR
APOSOR452038-1G
APOSOR452038-250MG
APOSOR452038-500MG
4-Chloro-DL-homophenylalanine 97%
4-Chloro-DL-homophenylalanine
Formula:
C₁₀H₁₂ClNO₂ MW: 213,661 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07372176 CAS Number: 225233-78-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...