2-(2-Chlorophenoxy)-benzoic acid 97%
Supplier: Apollo Scientific
OR452062-1G
OR452062-250MG
APOSOR452062-1GEA
568
EUR
APOSOR452062-1G
APOSOR452062-250MG
2-(2-Chlorophenoxy)-benzoic acid 97%
2-(2-Chlorophenoxy)-benzoic acid
Formula:
C₁₃H₉ClO₃ MW: 248,662 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06260646 CAS Number: 36809-08-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...