Fmoc-DL-norvaline 97%

Supplier: Apollo Scientific

OR452088-1G OR452088-25G OR452088-5G
APOSOR452088-1GEA 118 EUR
APOSOR452088-1G APOSOR452088-25G APOSOR452088-5G
Fmoc-DL-norvaline 97%
Fmoc-DL-norvaline
Formula: C₂₀H₂₁NO₄
MW: 339,385 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18263133
CAS Number: 144701-21-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR