Boc-N-methyl-D-norvaline 97%
Supplier: Apollo Scientific
OR452089-1G
OR452089-500MG
APOSOR452089-1GEA
394
EUR
APOSOR452089-1G
APOSOR452089-500MG
Boc-N-methyl-D-norvaline 97%
Boc-N-methyl-D-norvaline
Formula:
C₁₁H₂₁NO₄ MW: 231,289 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07366767 CAS Number: 177659-77-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...