5-(Cyclopropyl)thiophene-2-carboxylic acid
Supplier: Apollo Scientific
|
Warning
|
OR45211-1G
OR45211-250MG
OR45211-5G
APOSOR45211-1GEA
522
EUR
APOSOR45211-1G
APOSOR45211-250MG
APOSOR45211-5G
5-(Cyclopropyl)thiophene-2-carboxylic acid
5-(Cyclopropyl)thiophene-2-carboxylic acid
Formula:
C₈H₈O₂S MW: 168,213 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06802416 CAS Number: 29481-28-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...