1-Boc-2-acetyl-piperidine 97%
Supplier: Apollo Scientific
OR452124-1G
OR452124-500MG
APOSOR452124-1GEA
91.5
EUR
APOSOR452124-1G
APOSOR452124-500MG
1-Boc-2-acetyl-piperidine 97%
1-Boc-2-acetyl-piperidine
Formula:
C₁₂H₂₁NO₃ MW: 227,3 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09953375 CAS Number: 201991-24-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...