(R)-b-Amino-4-methyl-benzeneethanol 97%

Supplier: Apollo Scientific

Synonyms: (R)-2-Amino-2-(p-tolyl)ethanol

OR452141-1G OR452141-250MG
APOSOR452141-1GEA 358 EUR
APOSOR452141-1G APOSOR452141-250MG
(R)-b-Amino-4-methyl-benzeneethanol 97%
(R)-b-Amino-4-methyl-benzeneethanol
Formula: C₉H₁₃NO
MW: 151,206 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09253779
CAS Number: 639054-51-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR