(R)-b-Amino-cyclohexanepropanol 97%

Supplier: Apollo Scientific

OR452143-1G OR452143-250MG OR452143-500MG
APOSOR452143-1GEA 719 EUR
APOSOR452143-1G APOSOR452143-250MG APOSOR452143-500MG
(R)-b-Amino-cyclohexanepropanol 97%
(R)-b-Amino-cyclohexanepropanol
Formula: C₉H₁₉NO
Storage Temperature: Ambient
MDL Number: MFCD00672504
CAS Number: 205445-49-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR