b-Amino-4-methylbenzenepropanol 97%

Supplier: Apollo Scientific

Synonyms: 2-Amino-3-(p-tolyl)propan-1-ol

OR452173-1G OR452173-250MG
APOSOR452173-1GEA 789 EUR
APOSOR452173-1G APOSOR452173-250MG
b-Amino-4-methylbenzenepropanol 97%
b-Amino-4-methylbenzenepropanol
Formula: C₁₀H₁₅NO
MW: 165,232 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10034613
CAS Number: 35373-66-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR