1-(2-Nitrophenyl)ethanamine 97%
Supplier: Apollo Scientific
OR452222-1G
OR452222-500MG
APOSOR452222-1GEA
0
EUR
APOSOR452222-1G
APOSOR452222-500MG
1-(2-Nitrophenyl)ethanamine 97%
1-(2-Nitrophenyl)ethanamine
Formula:
C₈H₁₀N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD09041921 CAS Number: 100311-54-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...