1,1,1,2,3-Pentachloropropane

Supplier: Apollo Scientific
Warning

OR460069-100G OR460069-25G OR460069-5G
APOSOR460069-100GEA 1070 EUR
APOSOR460069-100G APOSOR460069-25G APOSOR460069-5G
1,1,1,2,3-Pentachloropropane
1,1,1,2,3-Pentachloropropane
Formula: C₃H₃Cl₅
MW: 216,321 g/mol
Boiling Pt: 72 °C (8 mm Hg)
Density: 1,61 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD01024425
CAS Number: 21700-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR