3-Bromo-5-iodophenol
Supplier: Apollo Scientific
|
Warning
|
OR46047-1G
OR46047-250MG
OR46047-5G
APOSOR46047-1GEA
123
EUR
APOSOR46047-1G
APOSOR46047-250MG
APOSOR46047-5G
3-Bromo-5-iodophenol
3-Bromo-5-iodophenol
Formula:
C₆H₄BrIO MW: 298,904 g/mol Melting Pt: 82…84 °C Storage Temperature: Ambient |
MDL Number:
MFCD12405397 CAS Number: 570391-20-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...