5-Bromopyrido[3,4-b]pyrazine

Supplier: Apollo Scientific
Warning

Synonyms: 5-Bromo-1,4,6-triazanaphthalene

OR46117-1G OR46117-250MG
APOSOR46117-1GEA 306 EUR
APOSOR46117-1G APOSOR46117-250MG
5-Bromopyrido[3,4-b]pyrazine
5-Bromopyrido[3,4-b]pyrazine
Formula: C₇H₄BrN₃
MW: 210,031 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12401005
CAS Number: 1159829-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR