3,5-Diiodopyridine

Supplier: Apollo Scientific

OR46151-1G OR46151-250MG
APOSOR46151-1GEA 238 EUR
APOSOR46151-1G APOSOR46151-250MG
3,5-Diiodopyridine
3,5-Diiodopyridine
Formula: C₅H₃I₂N
MW: 330,893 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01928488
CAS Number: 53710-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR