5-Bromo-2-iodophenol 98%

Supplier: Apollo Scientific
Warning

OR46220-10G OR46220-25G OR46220-5G
APOSOR46220-10GEA 236 EUR
APOSOR46220-10G APOSOR46220-25G APOSOR46220-5G
5-Bromo-2-iodophenol 98%
5-Bromo-2-iodophenol
Formula: C₆H₄BrIO
MW: 298,904 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16999931
CAS Number: 858855-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR