3,4-Dihydro-2H-pyran-5-boronic acid, pinacol ester
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-(3,4-Dihydro-2H-pyran-5-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydro-2H-pyran
OR46579-1G
OR46579-250MG
APOSOR46579-1GEA
764
EUR
APOSOR46579-1G
APOSOR46579-250MG
3,4-Dihydro-2H-pyran-5-boronic acid, pinacol ester
3,4-Dihydro-2H-pyran-5-boronic acid, pinacol ester
Formula:
C₁₁H₁₉BO₃ MW: 210,078 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD13195758 CAS Number: 1046811-99-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...