2-Amino-5-chlorobenzoyl chloride
Supplier: Apollo Scientific
OR46680-1G
OR46680-250MG
OR46680-500MG
APOSOR46680-1GEA
627
EUR
APOSOR46680-1G
APOSOR46680-250MG
APOSOR46680-500MG
2-Amino-5-chlorobenzoyl chloride
2-Amino-5-chlorobenzoyl chloride
Formula:
C₇H₅Cl₂NO MW: 190,027 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09901791 CAS Number: 910230-30-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...