2-Hydroxy-2-phenylacetyl chloride
Supplier: Apollo Scientific
|
Danger
|
OR46683-1G
OR46683-250MG
APOSOR46683-1GEA
510
EUR
APOSOR46683-1G
APOSOR46683-250MG
2-Hydroxy-2-phenylacetyl chloride
2-Hydroxy-2-phenylacetyl chloride
Formula:
C₈H₇ClO₂ MW: 170,593 g/mol Melting Pt: 116 °C Storage Temperature: Ambient |
MDL Number:
MFCD25963976 CAS Number: 50916-31-9 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...