3-Methyl-3,4-dihydro-2H-1,4-benzoxazine

Supplier: Apollo Scientific

Synonyms: 3-Methyl-3,4-dihydro-2H-benzo[b][1,4]oxazine

OR46715-1G OR46715-100MG OR46715-250MG OR46715-500MG OR46715-2.5G OR46715-5G
APOSOR46715-1GEA 418 EUR
APOSOR46715-1G APOSOR46715-100MG APOSOR46715-250MG APOSOR46715-500MG APOSOR46715-2.5G APOSOR46715-5G
3-Methyl-3,4-dihydro-2H-1,4-benzoxazine
3-Methyl-3,4-dihydro-2H-1,4-benzoxazine
Formula: C₉H₁₁NO
MW: 149.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01935996
CAS Number: 32329-20-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR