6-Cyclopropyl-2-sulphanylpyridine-3-carbonitrile

Supplier: Apollo Scientific
Warning

OR46755-1G OR46755-250MG
APOSOR46755-1GEA 157 EUR
APOSOR46755-1G APOSOR46755-250MG
6-Cyclopropyl-2-sulphanylpyridine-3-carbonitrile
6-Cyclopropyl-2-sulphanylpyridine-3-carbonitrile
Formula: C₉H₈N₂S
MW: 176,238 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19599987
CAS Number: 101045-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR