Methyl N-Cbz-2-piperidineacetate

Supplier: Apollo Scientific

OR470075-1G OR470075-5G
APOSOR470075-1GEA 306 EUR
APOSOR470075-1G APOSOR470075-5G
Methyl N-Cbz-2-piperidineacetate
Methyl N-Cbz-2-piperidineacetate
Formula: C₁₆H₂₁NO₄
MW: 291,342 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08461233
CAS Number: 169384-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR