4-Isobutyloxazolidine-2,5-dione

Supplier: Apollo Scientific

Synonyms: Isobutyloxazolidine-2,5-dione

OR470083-1G OR470083-5G
APOSOR470083-1GEA 121 EUR
APOSOR470083-1G APOSOR470083-5G
4-Isobutyloxazolidine-2,5-dione
4-Isobutyloxazolidine-2,5-dione
Formula: C₇H₁₁NO₃
MW: 157,167 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02684167
CAS Number: 51248-35-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR