Methyl 5-Bromo-2-hydroxy-3-propionylbenzoate
Supplier: Apollo Scientific
OR470180-1G
OR470180-5G
APOSOR470180-1GEA
275
EUR
APOSOR470180-1G
APOSOR470180-5G
Methyl 5-Bromo-2-hydroxy-3-propionylbenzoate
Methyl 5-Bromo-2-hydroxy-3-propionylbenzoate
Formula:
C₁₁H₁₁BrO₄ MW: 287,107 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01411506 CAS Number: 91099-82-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...