Ethyl 1-(Dimethylcarbamoyl)piperidine-2-carboxylate

Supplier: Apollo Scientific

OR470211-1G OR470211-5G
APOSOR470211-1GEA 194 EUR
APOSOR470211-1G APOSOR470211-5G
Ethyl 1-(Dimethylcarbamoyl)piperidine-2-carboxylate
Ethyl 1-(Dimethylcarbamoyl)piperidine-2-carboxylate
Formula: C₁₁H₂₀N₂O₃
MW: 228,288 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05695247
CAS Number: 1009341-34-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR