(S)-1-(Dimethylcarbamoyl)pyrrolidine-2-carboxylic acid

Supplier: Apollo Scientific

OR470215-1G OR470215-5G
APOSOR470215-1GEA 306 EUR
APOSOR470215-1G APOSOR470215-5G
(S)-1-(Dimethylcarbamoyl)pyrrolidine-2-carboxylic acid
(S)-1-(Dimethylcarbamoyl)pyrrolidine-2-carboxylic acid
Formula: C₈H₁₄N₂O₃
MW: 186,208 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14697611
CAS Number: 1305322-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR