3-(Hydroxymethyl)-5-(3-chlorophenyl)pyridine

Supplier: Apollo Scientific

OR470270-1G OR470270-5G
APOSOR470270-1GEA 601 EUR
APOSOR470270-1G APOSOR470270-5G
3-(Hydroxymethyl)-5-(3-chlorophenyl)pyridine
3-(Hydroxymethyl)-5-(3-chlorophenyl)pyridine
Formula: C₁₂H₁₀ClNO
Storage Temperature: Ambient
MDL Number: MFCD06410307
CAS Number: 887973-96-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR