Ethyl 1-Benzylimidazole-2-carboxylate
Supplier: Apollo Scientific
OR470295-25G
OR470295-5G
APOSOR470295-25GEA
1400
EUR
APOSOR470295-25G
APOSOR470295-5G
Ethyl 1-Benzylimidazole-2-carboxylate
Ethyl 1-Benzylimidazole-2-carboxylate
Formula:
C₁₃H₁₄N₂O₂ MW: 230,262 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11857953 CAS Number: 865998-45-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...