2-Amino-2-[4-(3-pyridyloxy)phenyl]acetic acid

Supplier: Apollo Scientific

OR470379-1G OR470379-5G
APOSOR470379-1GEA 469 EUR
APOSOR470379-1G APOSOR470379-5G
2-Amino-2-[4-(3-pyridyloxy)phenyl]acetic acid
2-Amino-2-[4-(3-pyridyloxy)phenyl]acetic acid
Formula: C₁₃H₁₂N₂O₃
MW: 244,246 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06410001
CAS Number: 1136884-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR