tert-Butyl N-Cbz-4-aminobutanoate

Supplier: Apollo Scientific

OR470585-1G OR470585-5G
APOSOR470585-1GEA 397 EUR
APOSOR470585-1G APOSOR470585-5G
tert-Butyl N-Cbz-4-aminobutanoate
tert-Butyl N-Cbz-4-aminobutanoate
Formula: C₁₆H₂₃NO₄
MW: 293,358 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22683325
CAS Number: 5105-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR