N-(3-Indolylmethyl)benzylamine

Supplier: Apollo Scientific

OR470605-1G OR470605-5G
APOSOR470605-1GEA 306 EUR
APOSOR470605-1G APOSOR470605-5G
N-(3-Indolylmethyl)benzylamine
N-(3-Indolylmethyl)benzylamine
Formula: C₁₆H₁₆N₂
MW: 236,312 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17430949
CAS Number: 57506-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR