2-(4-Thiazolyl)acetic acid hydrochloride
Supplier: Apollo Scientific
OR470621-1G
OR470621-5G
APOSOR470621-1GEA
306
EUR
APOSOR470621-1G
APOSOR470621-5G
2-(4-Thiazolyl)acetic acid hydrochloride
2-(4-Thiazolyl)acetic acid hydrochloride
Formula:
C₅H₅NO₂S·ClH MW: 179,625 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16295329 CAS Number: 1225286-62-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...