4-(Diethoxymethyl)-1-methylpiperidine
Supplier: Apollo Scientific
OR470629-1G
OR470629-5G
APOSOR470629-1GEA
194
EUR
APOSOR470629-1G
APOSOR470629-5G
4-(Diethoxymethyl)-1-methylpiperidine
4-(Diethoxymethyl)-1-methylpiperidine
Formula:
C₁₁H₂₃NO₂ MW: 201.306 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11554476 CAS Number: 1427475-31-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...