3-(Hydroxymethyl)-4-methylphenol

Supplier: Apollo Scientific
Warning

Synonyms: 5-Hydroxy-2-methylbenzyl alcohol

OR470760-5G OR470760-250MG OR470760-1G
APOSOR470760-5GEA 0 EUR
APOSOR470760-5G APOSOR470760-250MG APOSOR470760-1G
3-(Hydroxymethyl)-4-methylphenol
3-(Hydroxymethyl)-4-methylphenol
Formula: C₈H₁₀O₂
Storage Temperature: Ambient
MDL Number: MFCD18397253
CAS Number: 1261454-85-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR