3-Phenylpyrazin-2(1H)-one
Supplier: Apollo Scientific
OR470876-5G
OR470876-1G
APOSOR470876-5GEA
804
EUR
APOSOR470876-5G
APOSOR470876-1G
3-Phenylpyrazin-2(1H)-one
3-Phenylpyrazin-2(1H)-one
Formula:
C₁₀H₈N₂O MW: 172,183 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD24394632 CAS Number: 2882-18-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...