1-Oxo-1,2-dihydroisoquinoline-3-carboxylic acid

Supplier: Apollo Scientific

OR470943-1G OR470943-250MG
APOSOR470943-1GEA 804 EUR
APOSOR470943-1G APOSOR470943-250MG
1-Oxo-1,2-dihydroisoquinoline-3-carboxylic acid
1-Oxo-1,2-dihydroisoquinoline-3-carboxylic acid
Formula: C₁₀H₇NO₃
MW: 189,167 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01845162
CAS Number: 7509-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR