2-(2-Iodo-4-nitrophenyl)acetic acid
Supplier: Apollo Scientific
OR471044-1G
OR471044-250MG
APOSOR471044-1GEA
916
EUR
APOSOR471044-1G
APOSOR471044-250MG
2-(2-Iodo-4-nitrophenyl)acetic acid
2-(2-Iodo-4-nitrophenyl)acetic acid
Formula:
C₈H₆INO₄ Storage Temperature: Ambient |
MDL Number:
MFCD18398041 CAS Number: 66949-41-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...