Methyl 2-(Dimethoxyphosphoryl)propionate
Supplier: Apollo Scientific
OR471116-5G
OR471116-1G
APOSOR471116-5GEA
713
EUR
APOSOR471116-5G
APOSOR471116-1G
Methyl 2-(Dimethoxyphosphoryl)propionate
Methyl 2-(Dimethoxyphosphoryl)propionate
Formula:
C₆H₁₃O₅P MW: 196,138 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD21607028 CAS Number: 26530-60-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...