6-Phenylmorpholin-3-one
Supplier: Apollo Scientific
OR471184-1G
OR471184-250MG
APOSOR471184-1GEA
1010
EUR
APOSOR471184-1G
APOSOR471184-250MG
6-Phenylmorpholin-3-one
6-Phenylmorpholin-3-one
Formula:
C₁₀H₁₁NO₂ MW: 177,2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07781033 CAS Number: 5493-95-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...