Methyl 2-chloro-3-phenylpropionate 95%

Supplier: Apollo Scientific
Warning

Synonyms: Methyl 2-Chloro-3-phenylpropionate

OR471312-1G OR471312-250MG
APOSOR471312-1GEA 166 EUR
APOSOR471312-1G APOSOR471312-250MG
Methyl 2-chloro-3-phenylpropionate 95%
Methyl 2-chloro-3-phenylpropionate
Formula: C₁₀H₁₁ClO₂
MW: 198,646 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14538826
CAS Number: 18841-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR