1-Benzyl-6-oxopiperidine-3-carboxylic acid

Supplier: Apollo Scientific

OR471392-1G OR471392-5G OR471392-250MG
APOSOR471392-1GEA 397 EUR
APOSOR471392-1G APOSOR471392-5G APOSOR471392-250MG
1-Benzyl-6-oxopiperidine-3-carboxylic acid
1-Benzyl-6-oxopiperidine-3-carboxylic acid
Formula: C₁₃H₁₅NO₃
MW: 233.263 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17041934
CAS Number: 32749-55-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR