Methyl N-Cbz-3-piperidylacetate

Supplier: Apollo Scientific

OR471455-1G OR471455-5G
APOSOR471455-1GEA 256 EUR
APOSOR471455-1G APOSOR471455-5G
Methyl N-Cbz-3-piperidylacetate
Methyl N-Cbz-3-piperidylacetate
Formula: C₁₆H₂₁NO₄
MW: 291,342 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02179007
CAS Number: 86827-08-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR