4-Chloro-7-hydroxyquinazoline

Supplier: Apollo Scientific

OR471464-250MG OR471464-100MG
APOSOR471464-250MGEA 877 EUR
APOSOR471464-250MG APOSOR471464-100MG
4-Chloro-7-hydroxyquinazoline
4-Chloro-7-hydroxyquinazoline
Formula: C₈H₅ClN₂O
MW: 180,591 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08276177
CAS Number: 849345-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR