6-Chloro-9-methyl-7H-purin-8(9H)-one

Supplier: Apollo Scientific
Danger

OR471472-1G OR471472-250MG OR471472-5G
APOSOR471472-1GEA 713 EUR
APOSOR471472-1G APOSOR471472-250MG APOSOR471472-5G
6-Chloro-9-methyl-7H-purin-8(9H)-one
6-Chloro-9-methyl-7H-purin-8(9H)-one
Formula: C₆H₅ClN₄O
MW: 184,583 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22207346
CAS Number: 84602-79-9
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR