2-Chloro-9-methyl-7H-purin-8(9H)-one

Supplier: Apollo Scientific

OR471476-1G OR471476-250MG
APOSOR471476-1GEA 804 EUR
APOSOR471476-1G APOSOR471476-250MG
2-Chloro-9-methyl-7H-purin-8(9H)-one
2-Chloro-9-methyl-7H-purin-8(9H)-one
Formula: C₆H₅ClN₄O
MW: 184,583 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24567184
CAS Number: 1273315-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR