Methyl 3,4-Dichloropyridine-2-carboxylate
Supplier: Apollo Scientific
OR471484-1G
OR471484-250MG
APOSOR471484-1GEA
397
EUR
APOSOR471484-1G
APOSOR471484-250MG
Methyl 3,4-Dichloropyridine-2-carboxylate
Methyl 3,4-Dichloropyridine-2-carboxylate
Formula:
C₇H₅Cl₂NO₂ MW: 206,026 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16658921 CAS Number: 343781-52-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...