N2-Boc-3-methoxy-1,2-benzenediamine

Supplier: Apollo Scientific

OR471507-1G OR471507-250MG OR471507-5G
APOSOR471507-1GEA 601 EUR
APOSOR471507-1G APOSOR471507-250MG APOSOR471507-5G
N2-Boc-3-methoxy-1,2-benzenediamine
N2-Boc-3-methoxy-1,2-benzenediamine
Formula: C₁₂H₁₈N₂O₃
MW: 238,283 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09701243
CAS Number: 954238-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR