N1-Boc-5-methoxy-1,2-benzenediamine

Supplier: Apollo Scientific
Warning

OR471532-1G OR471532-250MG
APOSOR471532-1GEA 284 EUR
APOSOR471532-1G APOSOR471532-250MG
N1-Boc-5-methoxy-1,2-benzenediamine
N1-Boc-5-methoxy-1,2-benzenediamine
Formula: C₁₂H₁₈N₂O₃
MW: 238,283 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09701255
CAS Number: 362670-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR